Skip to Content
Merck
All Photos(1)

Key Documents

304220

Sigma-Aldrich

4-Bromo-2-fluoroaniline

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(F)NH2
CAS Number:
Molecular Weight:
190.01
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

40-42 °C

functional group

bromo

SMILES string

Nc1ccc(Br)cc1F

InChI

1S/C6H5BrFN/c7-4-1-2-6(9)5(8)3-4/h1-3H,9H2

InChI key

GZRMNMGWNKSANY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Bromo-2-fluoroaniline was used in synthesis of 4-amino-3-fluorophenyl boronic acid. It was also used in chemical modification and synthesis of boronic acid derivatives for ultraviolet-visible titration.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Synthesis and crystal structure of 4-amino-3-fluorophenylboronic acid.
Das S, et al.
Tetrahedron Letters, 44(42), 7719-7722 (2003)
Vladimir L Alexeev et al.
Clinical chemistry, 50(12), 2353-2360 (2004-10-02)
We recently developed a photonic crystal glucose-sensing material, which consists of a crystalline colloidal array embedded within a polymer network of a polyacrylamide-poly(ethylene glycol) hydrogel with pendent phenylboronic acid groups. The aim of the present work was to improve this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service