Skip to Content
Merck
All Photos(1)

Key Documents

299340

Sigma-Aldrich

Glycine-1-13C,15N

99 atom % 13C, 98 atom % 15N

Synonym(s):

13C and 15N Labeled aminoacetic acid, 13C and 15N Labeled glycine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H215NCH213CO2H
CAS Number:
Molecular Weight:
77.05
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C
98 atom % 15N

Quality Level

form

solid

mp

240 °C (dec.) (lit.)

mass shift

M+2

SMILES string

[15NH2]C[13C](O)=O

InChI

1S/C2H5NO2/c3-1-2(4)5/h1,3H2,(H,4,5)/i2+1,3+1

InChI key

DHMQDGOQFOQNFH-SUEIGJEOSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julian P Maier et al.
Cell death & disease, 12(8), 723-723 (2021-07-23)
Glioblastoma (GBM), the most malignant tumor of the central nervous system, is marked by its dynamic response to microenvironmental niches. In particular, this cellular plasticity contributes to the development of an immediate resistance during tumor treatment. Novel insights into the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service