Skip to Content
Merck
All Photos(4)

Key Documents

299286

Sigma-Aldrich

Ammonium-15N2 sulfate

98 atom % 15N

Synonym(s):

15N Labeled ammonium sulfate, Diazanium-15N2 sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(15NH4)2SO4
CAS Number:
Molecular Weight:
134.13
MDL number:
UNSPSC Code:
12142200
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % 15N

form

solid

technique(s)

bio NMR: suitable
protein expression: suitable

mp

>280 °C (dec.) (lit.)

mass shift

M+2

SMILES string

[15NH3].[15NH3].OS(O)(=O)=O

InChI

1S/2H3N.H2O4S/c;;1-5(2,3)4/h2*1H3;(H2,1,2,3,4)/i2*1+1;

InChI key

BFNBIHQBYMNNAN-ISOLYIDJSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ammonium-15N2 sulfate can be used:
  • In the quantitative protein expression profiling.
  • In metabolomic analyses to identify the number of nitrogen atoms in each metabolite and to confirm biological origin.
  • To determine the agronomic fertilizer nitrogen absorption efficiency by the coffee plant.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R T Batey et al.
Nucleic acids research, 20(17), 4515-4523 (1992-09-11)
A general method for large scale preparation of uniformly isotopically labeled ribonucleotides and RNAs is described. Bacteria are grown on isotopic growth medium, and their nucleic acids are harvested and degraded to mononucleotides. These are enzymatically converted into ribonucleoside triphosphates
Energy Nucl. Agric., 9, 66-66 (1988)
Izargi Vega-Mas et al.
Scientific reports, 9(1), 8925-8925 (2019-06-22)
Proper carbon (C) supply is essential for nitrogen (N) assimilation especially when plants are grown under ammonium (NH4+) nutrition. However, how C and N metabolic fluxes adapt to achieve so remains uncertain. In this work, roots of wheat (Triticum aestivum

Articles

Utilizing ISOGRO® Supplementation of M9 Minimal Media to Enhance Recombinant Protein Expression.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service