Skip to Content
Merck
All Photos(2)

Key Documents

298964

Sigma-Aldrich

N,N′-Diisopropylethylenediamine

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHNHCH2CH2NHCH(CH3)2
CAS Number:
Molecular Weight:
144.26
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

refractive index

n20/D 1.4289 (lit.)

bp

169-171 °C (lit.)

density

0.798 g/mL at 25 °C (lit.)

SMILES string

CC(C)NCCNC(C)C

InChI

1S/C8H20N2/c1-7(2)9-5-6-10-8(3)4/h7-10H,5-6H2,1-4H3

InChI key

MFIGJRRHGZYPDD-UHFFFAOYSA-N

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chenshu Liu et al.
Experimental and therapeutic medicine, 18(3), 1669-1676 (2019-08-15)
Previous studies examining whether autophagy has a protective or deleterious role during ischemia/reperfusion (I/R) injury have reported a varying role in different organs and remains a matter of debate. The aim of the current study was to explore the role
Jesús García-Valdés et al.
Journal of inorganic biochemistry, 197, 110689-110689 (2019-05-18)
Bis-cations with two 2,3-diferrocenylcyclopropenium fragments 3a-d, and the cis-2-(1,2-diferrocenylvinyl)-2-imidazolinium tetrafluoroborates 4a, d or the cis-2-(1,2-diferrocenylvinyl)-3,4,5,6-tetrahydropyrimidin-2-ium tetrafluoroborates 4b, c were obtained by interactions of 2,3-diferrocenyl-1-ethoxycyclopropenium tetrafluoroborate 1 with bis-1,4-N,N-(2a, d) or bis-1,5-N,N-(2b, c) nucleophiles. The reactions of 3a-d with sodium azide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service