29823
cis-4-Cyclopentene-1,3-diol
≥99.0% (GC)
Synonym(s):
cis-3,5-Dihydroxy-1-cyclopentene
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C5H8O2
CAS Number:
Molecular Weight:
100.12
Beilstein:
2039395
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥99.0% (GC)
form
solid
mp
55-59 °C
functional group
hydroxyl
SMILES string
O[C@H]1C[C@@H](O)C=C1
InChI
1S/C5H8O2/c6-4-1-2-5(7)3-4/h1-2,4-7H,3H2/t4-,5+
InChI key
IGRLIBJHDBWKNA-SYDPRGILSA-N
Application
cis-4-Cyclopentene-1,3-diol is an important building block for the synthesis of natural products with cyclopentanoid structure elements. It was used in preparation of key intermediates of prostaglandin synthesis.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
M. Sato et al.
Chemistry Letters (Jpn), 1057-1057 (1985)
R. Noyori et al.
Angewandte Chemie (International Edition in English), 96, 854-854 (1984)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service