Skip to Content
Merck
All Photos(1)

Key Documents

295906

Sigma-Aldrich

Poly(ethylene glycol)

average Mn 2,050, chips

Synonym(s):

Polyethylene glycol, PEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.23

product name

Poly(ethylene glycol), average Mn 2,050, chips

form

chips

Quality Level

mol wt

average Mn 2,050

mp

52-54 °C

Ω-end

hydroxyl

α-end

hydroxyl

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Cytotoxicity Study of Polyethylene Glycol Derivatives: Evaluates the cytotoxic effects of various PEG derivatives, important for chemists in pharmaceutical development and safety assessment (Liu et al., 2017).
  • ExtraPEG: A Polyethylene Glycol-Based Method for Enrichment of Extracellular Vesicles: Introduces a PEG-based method for isolating exosomes, useful for researchers in biomedical and clinical sciences (Rider et al., 2016).

Other Notes

Molecular weight: Mn 1,900-2,200

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paras R Patel et al.
Journal of neural engineering, 12(4), 046009-046009 (2015-06-03)
Single carbon fiber electrodes (d = 8.4 μm) insulated with parylene-c and functionalized with pTS have been shown to record single unit activity but manual implantation of these devices with forceps can be difficult. Without an improvement in the insertion
Elissa J Welle et al.
Journal of neural engineering, 17(2), 026037-026037 (2020-03-27)
Carbon fiber electrodes may enable better long-term brain implants, minimizing the tissue response commonly seen with silicon-based electrodes. The small diameter fiber may enable high-channel count brain-machine interfaces capable of reproducing dexterous movements. Past carbon fiber electrodes exhibited both high
Chenlu Zhang et al.
PloS one, 12(9), e0184730-e0184730 (2017-09-09)
The economic production of cellulosic biofuel requires efficient and full utilization of all abundant carbohydrates naturally released from plant biomass by enzyme cocktails. Recently, we reconstituted the Neurospora crassa xylodextrin transport and consumption system in Saccharomyces cerevisiae, enabling growth of
Paras R Patel et al.
Journal of neural engineering, 17(5), 056029-056029 (2020-10-16)
Multimodal measurements at the neuronal level allow for detailed insight into local circuit function. However, most behavioral studies focus on one or two modalities and are generally limited by the available technology. Here, we show a combined approach of electrophysiology
Idalis Villanueva et al.
Acta biomaterialia, 5(8), 2832-2846 (2009-06-11)
The pericellular matrix (PCM) surrounding chondrocytes is thought to play an important role in transmitting biochemical and biomechanical signals to the cells, which regulates many cellular functions including tissue homeostasis. To better understand chondrocytes interactions with their PCM, three-dimensional poly(ethylene

Articles

Click chemistry, and the copper(I)-catalyzed azide-alkyne cycloaddition (CuAAC) in particular, is a powerful new synthetic tool in polymer chemistry and material science.

Click chemistry, and the copper(I)-catalyzed azide-alkyne cycloaddition (CuAAC) in particular, is a powerful new synthetic tool in polymer chemistry and material science.

Click chemistry, and the copper(I)-catalyzed azide-alkyne cycloaddition (CuAAC) in particular, is a powerful new synthetic tool in polymer chemistry and material science.

Click chemistry, and the copper(I)-catalyzed azide-alkyne cycloaddition (CuAAC) in particular, is a powerful new synthetic tool in polymer chemistry and material science.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service