All Photos(1)
About This Item
Linear Formula:
C2H5CH=C(CH3)CHO
CAS Number:
Molecular Weight:
98.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
refractive index
n20/D 1.45 (lit.)
bp
137-138 °C/765 mmHg (lit.)
density
0.86 g/mL at 25 °C (lit.)
functional group
aldehyde
SMILES string
[H]C(=O)\C(C)=C\CC
InChI
1S/C6H10O/c1-3-4-6(2)5-7/h4-5H,3H2,1-2H3/b6-4+
InChI key
IDEYZABHVQLHAF-GQCTYLIASA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Hydrodeoxygenation and hydrogenation of 2-methyl-2-pentenal on supported platinum, palladium and copper catalysts has been reported.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 3 - Skin Sens. 1
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
89.6 °F - closed cup
Flash Point(C)
32 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Hydrogenation and Hydrodeoxygenation of 2-methyl-2-pentenal on supported metal catalysts.
Pham TT, et al.
J. Catal., 266(1), 9-14 (2009)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service