Skip to Content
Merck
All Photos(1)

Key Documents

290580

Sigma-Aldrich

2-Phenethyl acetate

99%

Synonym(s):

2-Phenylethyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH2CH2C6H5
CAS Number:
Molecular Weight:
164.20
Beilstein:
638179
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

5.67 (vs air)

Assay

99%

form

liquid

refractive index

n20/D 1.498 (lit.)

bp

238-239 °C (lit.)

density

1.032 g/mL at 25 °C (lit.)

functional group

ester
phenyl

SMILES string

CC(=O)OCCc1ccccc1

InChI

1S/C10H12O2/c1-9(11)12-8-7-10-5-3-2-4-6-10/h2-6H,7-8H2,1H3

InChI key

MDHYEMXUFSJLGV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Phenethyl acetate is a highly valued natural volatile ester with a rose-like odour and is widely used to add scent or flavour to cosmetics, soaps, foods and drinks.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

221.0 °F - closed cup

Flash Point(C)

105 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elda Vitanović et al.
Journal of economic entomology, 113(2), 752-759 (2019-12-28)
The olive fruit fly, Bactrocera oleae (Rossi), is one of the most damaging insect pests of olives worldwide, requiring the use of insecticides for fruit protection in many orchards. Olive fruit flies are attracted to volatile composunds, including a female-produced
Zuobing Xiao et al.
Food research international (Ottawa, Ont.), 116, 211-222 (2019-02-06)
This study focused on the impact of esters on the perception of floral aroma in rose essential oil. Various aromatic reconstitutions were prepared, consisting of 10 alcohols and 9 esters, all the concentrations found in rose essential oil. Sensory analysis
Philipp Adler et al.
Enzyme and microbial technology, 48(3), 285-292 (2011-11-25)
An unstructured model for an integrated fermentation/membrane extraction process for the production of the aroma compounds 2-phenylethanol and 2-phenylethylacetate by Kluyveromyces marxianus CBS 600 was developed. The extent to which this model, based only on data from the conventional fermentation
Patrick C Aculey et al.
Journal of food science, 75(6), S300-S307 (2010-08-21)
Export of cocoa beans is of great economic importance in Ghana and several other tropical countries. Raw cocoa has an astringent, unpleasant taste, and flavor, and has to be fermented, dried, and roasted to obtain the characteristic cocoa flavor and
Fernando Viana et al.
International journal of food microbiology, 151(2), 235-240 (2011-10-04)
The effect of simultaneous or sequential inoculation of Hanseniaspora vineae CECT 1471 and Saccharomyces cerevisiae T73 in non-sterile must on 2-phenylethyl acetate production has been examined. In both treatments tested, no significant differences in Saccharomyces yeast growth were found, whereas

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service