Skip to Content
Merck
All Photos(3)

Key Documents

288640

Sigma-Aldrich

Manganese(II) nitrate hydrate

98%

Synonym(s):

Manganese(2+) dinitrate hydrate, Manganous dinitrate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mn(NO3)2·xH2O
CAS Number:
Molecular Weight:
178.95 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
solid

grade

for analytical purposes

Assay

98%

form

solid

composition

Degree of hydration, 4-6

storage temp.

2-8°C

SMILES string

O.[Mn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Mn.2NO3.H2O/c;2*2-1(3)4;/h;;;1H2/q+2;2*-1;

InChI key

HBTFASPVVFSRRI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Manganese(II) nitrate hydrate is an inorganic salt highly soluble in water. It can be used as a source of manganese ions in the synthesis of various manganese-containing materials for batteries and supercapacitor applications. Additionally, it also serves as a safe, cost-effective, and efficient electrolyte for rapid energy storage applications.

Application

Manganese(II) nitrate hydrate can be used as:      
  • A precursor for the synthesis of manganese dioxide (MnO2), which is a common cathode material in primary alkaline batteries. 
  • A precursor in the synthesis of manganese dioxide (MnO2) for use in flexible micro supercapacitors. MnO2 exhibits high specific capacitance and excellent cycling stability, making it suitable for rapid energy storage applications.    
  • A manganese source in the colloidal solution combustion synthesis process to produce 3D δ-MnO2 nanostructures with ultra-large mesopores used in the fabrication of high-performance lithium-ion battery anodes.     
  • A precursor in the microwave-aided fabrication of calcium-substituted dysprosium perovskite manganite oxide nanocomposites (DyMnO3) for supercapacitor applications.

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Separation of rare earths from transition metals by liquid-liquid extraction from a molten salt hydrate to an ionic liquid phase
Rout A and Binnemans K
Dalton Transactions, 43(8), 3186-3195 (2014)
Reactive ceramics of CeO2-MOx (M= Mn, Fe, Ni, Cu) for H2 generation by two-step water splitting using concentrated solar thermal energy
Kaneko H, et al.
Energy, 32(5), 656-663 (2007)
Thermal characteristics of manganese (II) nitrate hexahydrate as a phase change material for cooling systems
Nagano K, et al.
Applied Thermal Engineering, 23(2), 229-241 (2003)
Jaechan Ryu et al.
Nature communications, 9(1), 3715-3715 (2018-09-15)
Aluminum-air batteries are promising candidates for next-generation high-energy-density storage, but the inherent limitations hinder their practical use. Here, we show that silver nanoparticle-mediated silver manganate nanoplates are a highly active and chemically stable catalyst for oxygen reduction in alkaline media.
Breno R Barrioni et al.
Journal of colloid and interface science, 547, 382-392 (2019-04-12)
Bioactive glass nanoparticles (BGNPs) are of great interest in tissue engineering as they possess high dissolution rate and capability of being internalized by cells, releasing their dissolution products with therapeutic benefits intracellularly. A modified Stöber process can be applied to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service