Skip to Content
Merck
All Photos(2)

Key Documents

288020

Sigma-Aldrich

Hexamethylditin

99%

Synonym(s):

Hexamethyldistannane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SnSn(CH3)3
CAS Number:
Molecular Weight:
327.63
Beilstein:
3600327
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

99%

bp

182 °C/756 mmHg (lit.)

mp

23-24 °C (lit.)

density

1.58 g/mL at 20 °C (lit.)

SMILES string

C[Sn](C)(C)[Sn](C)(C)C

InChI

1S/6CH3.2Sn/h6*1H3;;

InChI key

CCRMAATUKBYMPA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent used to prepare aryl tin compounds for microwave-assisted Stille cross-coupling with halopyridines[1] or copper-mediated O-arylation of phenols.[2]

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

141.8 °F - closed cup

Flash Point(C)

61 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 47, 8607-8607 (2006)
Tetrahedron Letters, 47, 8973-8973 (2006)
Marcus Unverricht-Yeboah et al.
Journal of radiation research, 59(4), 411-429 (2018-05-26)
Gene expression analysis was carried out in Jurkat cells in order to identify candidate genes showing significant gene expression alterations allowing robust discrimination of the Auger emitter 123I, incorporated into the DNA as 123I-iododeoxyuridine (123IUdR), from α- and γ-radiation. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service