Skip to Content
Merck
All Photos(1)

Key Documents

286052

Sigma-Aldrich

Heneicosane

98%

Synonym(s):

n-Heneicosane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)19CH3
CAS Number:
Molecular Weight:
296.57
Beilstein:
1748500
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

10.3 (vs air)

Quality Level

vapor pressure

<1 mmHg ( 20 °C)

Assay

98%

form

solid

bp

100 °C/2 mmHg (lit.)
356.1 °C/760 mmHg

mp

39-41 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCCCCC

InChI

1S/C21H44/c1-3-5-7-9-11-13-15-17-19-21-20-18-16-14-12-10-8-6-4-2/h3-21H2,1-2H3

InChI key

FNAZRRHPUDJQCJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Heneicosane is one of the important components of Carthamus tinctorius flowers essential oil, essential oil from Tragopogon graminifolius DC and essential oils from the bud of Citrus aurantium L. var. amara Engl.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinous Asgarpanah et al.
Chinese journal of integrative medicine, 19(2), 153-159 (2013-02-02)
Carthamus tinctorius L. is commonly known as Safflower. C. tinctorius extracts and oil are important in drug development with numerous pharmacological activities in the world. This plant is cultivated mainly for its seed, which is used as edible oil. For
Mohammad Hosein Farzaei et al.
Natural product communications, 9(1), 121-124 (2014-03-26)
Tragopogon graminifolius DC., family Compositae, is widely consumed as a green vegetable in the west of Iran and for the treatment of gastrointestinal and hepatic ailments. In this study, the chemical composition of the essential oil from T. graminifolius aerial
Ming-Hua Jiang et al.
Journal of food science, 76(9), C1219-C1225 (2012-03-16)
A comparative study of steam distillation extraction (SDE), reflux extraction (RE), and ultrasound-assisted extraction (USE) was conducted for the extraction of essential oils from the bud of Citrus aurantium L. var. amara Engl. Each method was evaluated in terms of
Gazendra Shakya et al.
Science advances, 6(14), eaaz7188-eaaz7188 (2020-04-15)
Liquid emulsion droplet evaporation is of importance for various sensing and imaging applications. The liquid-to-gas phase transformation is typically triggered thermally or acoustically by low-boiling point liquids, or by inclusion of solid structures that pin the vapor/liquid contact line to
Luana Quassinti et al.
Fitoterapia, 97, 133-141 (2014-06-14)
Smyrnium olusatrum (Apiaceae), well known as wild celery, is a biennal celery-scented plant used for many centuries as a vegetable, then abandoned after the introduction of celery. In the present work, the essential oil obtained from inflorescences and the amounts

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service