Skip to Content
Merck
All Photos(2)

Key Documents

284777

Sigma-Aldrich

Methyl 6-methylpyridine-3-carboxylate

97%

Synonym(s):

Methyl 6-methylnicotinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9NO2
CAS Number:
Molecular Weight:
151.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

bp

160 °C/106 mmHg (lit.)

mp

34-37 °C (lit.)

functional group

ester

SMILES string

COC(=O)c1ccc(C)nc1

InChI

1S/C8H9NO2/c1-6-3-4-7(5-9-6)8(10)11-2/h3-5H,1-2H3

InChI key

VYPPZXZHYDSBSJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methyl 6-methylpyridine-3-carboxylate may be used in chemical synthesis studies.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

217.4 °F - closed cup

Flash Point(C)

103 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service