Skip to Content
Merck
All Photos(1)

Key Documents

283835

Sigma-Aldrich

Bromoacetic acid-13C2

99 atom % 13C

Synonym(s):

13C Labeled bromoacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br13CH213CO2H
CAS Number:
Molecular Weight:
140.93
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C

Assay

99% (CP)

form

powder

technique(s)

protein expression: suitable

bp

208 °C (lit.)

mp

47-49 °C (lit.)

mass shift

M+2

SMILES string

O[13C](=O)[13CH2]Br

InChI

1S/C2H3BrO2/c3-1-2(4)5/h1H2,(H,4,5)/i1+1,2+1

InChI key

KDPAWGWELVVRCH-ZDOIIHCHSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1A - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service