All Photos(1)
About This Item
Linear Formula:
H2N13CH213CO2H
CAS Number:
Molecular Weight:
77.05
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.12
Recommended Products
isotopic purity
99 atom % 13C
form
powder
mp
240 °C (dec.) (lit.)
mass shift
M+2
SMILES string
N[13CH2][13C](O)=O
InChI
1S/C2H5NO2/c3-1-2(4)5/h1,3H2,(H,4,5)/i1+1,2+1
InChI key
DHMQDGOQFOQNFH-ZDOIIHCHSA-N
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Wenfan Ke et al.
Nature communications, 11(1), 2587-2587 (2020-05-24)
The gut microbiota metabolizes drugs and alters their efficacy and toxicity. Diet alters drugs, the metabolism of the microbiota, and the host. However, whether diet-triggered metabolic changes in the microbiota can alter drug responses in the host has been largely
Andrea E Steuer et al.
Metabolites, 10(8) (2020-07-31)
Psychoactive stimulants are a popular drug class which are used recreationally. Over the last decade, large numbers of new psychoactive substances (NPS) have entered the drug market and these pose a worldwide problem to human health. Metabolomics approaches are useful
Volker Döring et al.
ACS synthetic biology, 7(9), 2029-2036 (2018-08-15)
Endowing biotechnological platform organisms with new carbon assimilation pathways is a key challenge for industrial biotechnology. Here we report progress toward the construction of formatotrophic Escherichia coli strains. Glycine and serine, universal precursors of one-carbon compounds oxidized during heterotrophic growth
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service