281883
Potassium trifluoroacetate
98%
Synonym(s):
Trifluoroacetic acid potassium salt
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
98%
mp
140-142 °C (lit.)
density
1.49 g/mL (lit.)
functional group
fluoro
SMILES string
[K+].[O-]C(=O)C(F)(F)F
InChI
1S/C2HF3O2.K/c3-2(4,5)1(6)7;/h(H,6,7);/q;+1/p-1
InChI key
CUNPJFGIODEJLQ-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
General description
Potassium trifluoroacetate is used as a versatile chemical reagent in various organic reactions. It can be employed as a source of trifluoroacetyl anion (CF3CO2-) in reactions involving nucleophilic substitution and acylation reactions.
Application
Potassium trifluoroacetate can be used as a reagent in trifluoromethylation reactions. It is used to synthesize trifluoromethyl thioethers via iron(II) chloride-catalyzed decarboxylation as well as a subsequent trifluoromethylation of organothiocyanates.
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
ACS nano, 9(2), 1995-2008 (2015-01-27)
The development of well-defined polymeric nanoparticles (NPs) as delivery carriers for antimicrobials targeting human infectious diseases requires rational design of the polymer template, an efficient synthetic approach, and fundamental understanding of the developed NPs, e.g., drug loading/release, particle stability, and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service