Skip to Content
Merck
All Photos(2)

Key Documents

281832

Sigma-Aldrich

Mesityl oxide

technical grade, 90%

Synonym(s):

4-Methyl-3-penten-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCOCH3
CAS Number:
Molecular Weight:
98.14
Beilstein:
1361550
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

Assay

90%

form

liquid

refractive index

n20/D 1.442 (lit.)

bp

130 °C (lit.)

mp

-41.5 °C (lit.)

density

0.858 g/mL at 25 °C (lit.)

functional group

ketone

SMILES string

C\C(C)=C\C(C)=O

InChI

1S/C6H10O/c1-5(2)4-6(3)7/h4H,1-3H3

InChI key

SHOJXDKTYKFBRD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Zinc chloride-catalyzed condensation of mesityl oxide with ethyl acetoacetate has been reported.

Application

Mesityl oxide has been used:
  • in the synthesis of 2,3,4,5-tetrahydro-1,3,3-trimethyldipyrrin, a crucial building block in the rational synthesis of chlorins and oxochlorins
  • as neutral marker in the preparation of imidazole-coated capillary column for electrophoresis

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

87.8 °F - closed cup

Flash Point(C)

31 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marcin Ptaszek et al.
Organic process research & development, 9(5), 651-659 (2005-01-01)
2,3,4,5-Tetrahydro-1,3,3-trimethyldipyrrin (1) is a crucial building block in the rational synthesis of chlorins and oxochlorins. The prior 5-step synthesis of 1 from pyrrole-2-carboxaldehyde (2) employed relatively simple and well-known reactions yet suffered from several drawbacks, including limited scale (>/= 0.5
C Y Liu et al.
Journal of chromatography. A, 897(1-2), 383-392 (2000-12-29)
An imidazole-coated capillary column for electrophoresis has been prepared by means of organosilanization. With mesityl oxide as neutral marker, the results indicated that the electroosmotic flow of the bonded phase displays a dramatic difference in pH dependence in comparison with
Condensation of mesityl oxide with acetoacetic ester.
Surmatis JD, et al.
The Journal of Organic Chemistry, 35(4), 1053-1056 (1970)
Christian Starkenmann
Journal of agricultural and food chemistry, 51(24), 7146-7155 (2003-11-13)
Cysteine conjugates, resulting from the addition of cysteine to alpha,beta-unsaturated carbonyl compounds, are important precursors of odorant sulfur compounds in food flavors. The aim of this work was to better understand this chemistry in the light of the unexpected double
Scott E Denmark et al.
The Journal of organic chemistry, 71(4), 1668-1676 (2006-02-14)
The mechanism of the formation of substituted quinolines from anilines and alpha,beta-unsaturated ketones has been studied by the use of 13C-labeled ketones in cross-over experiments. In the reaction of doubly labeled 13C(2,4) mesityl oxide, a 100% scrambling of the label

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service