Skip to Content
Merck
All Photos(1)

Key Documents

280550

Sigma-Aldrich

4-Diphenylmethoxy-1-methylpiperidine hydrochloride

95%

Synonym(s):

4-Diphenylmethoxy-1-methylpiperidine, Diphenylpyraline hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H23NO · HCl
CAS Number:
Molecular Weight:
317.85
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

mp

203-205 °C (lit.)

functional group

ether
phenyl

SMILES string

Cl[H].CN1CCC(CC1)OC(c2ccccc2)c3ccccc3

InChI

1S/C19H23NO.ClH/c1-20-14-12-18(13-15-20)21-19(16-8-4-2-5-9-16)17-10-6-3-7-11-17;/h2-11,18-19H,12-15H2,1H3;1H

InChI key

LPRLDRXGWKXRMQ-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

General description

4-Diphenylmethoxy-1-methylpiperidine hydrochloride is also referred as diphenylpyraline hydrochloride (DPP), is a first-generation antihistamine.

Application

4-Diphenylmethoxy-1-methylpiperidine hydrochloride was used as a pharmacotherapy for Parkinson′s disease.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Ohno et al.
Journal of the neurological sciences, 182(2), 95-97 (2001-01-04)
A long-term follow-up study was made of three patients with cerebrotendinous xanthomatosis (CTX) associated with parkinsonism, two of whom were siblings. Besides typical CTX symptoms, all three patients showed severe parkinsonism. This observation has been rarely reported in CTX. The
K O Ebete et al.
Journal of chromatography. B, Biomedical applications, 677(2), 319-323 (1996-03-03)
A rapid, reliable and specific reversed-phase high-performance liquid chromatographic procedure is described for the determination of diphenylpyraline hydrochloride at nanogram concentrations in plasma and urine. After extraction of the drug with n-pentane-2-propanol (50:1) from alkalinized samples, the organic extract was
N Shibata et al.
Rinsho shinkeigaku = Clinical neurology, 30(9), 978-984 (1990-09-01)
Two sibling cases of cerebrotendinous xanthomatosis with parkinsonism were reported. One was a woman of 39 years old, and another was her sister of 36 years old. In both cases, febrile convulsion appeared on 1.5 year old, and mental deterioration
Gennady B Lapa et al.
European journal of pharmacology, 506(3), 237-240 (2005-01-04)
Diphenylpyraline hydrochloride (DPP) is used clinically as an antihistamine drug, but its neurobiological effects are not completely understood. Voltammetry and microdialysis were used to investigate potential actions of DPP on the dopamine system. Voltammetric monitoring of dopamine signals in mouse
M A Lung et al.
Rhinology, 25(2), 95-100 (1987-06-01)
The effects of three commonly used H1 antihistamines on the nasal vascular and airway resistances were studied in the dog. Promethazine hydrochloride decreased nasal vascular resistance but increased nasal airway resistance in a dose-dependent manner. Diphenylpyraline hydrochloride in low doses

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service