Skip to Content
Merck
All Photos(1)

Key Documents

276332

Sigma-Aldrich

Rubidium

ingot, 99.6% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Rb
CAS Number:
Molecular Weight:
85.47
EC Number:
MDL number:
UNSPSC Code:
11101711
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.6% trace metals basis

form

ingot

reaction suitability

reagent type: reductant

packaging

pkg of Packaged in: Breakseal Ampule

resistivity

11.0 μΩ-cm, 20°C

impurities

0.2-0.4% Cs

bp

686 °C (lit.)

mp

38-39 °C (lit.)

density

1.53 g/mL at 25 °C (lit.)

SMILES string

[Rb]

InChI

1S/Rb

InChI key

IGLNJRXAVVLDKE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Rubidium is a highly electropositive alkaline metal that can be obtained as a byproduct of refining lithium from lepidolite. It is used in photomultiplier tubes, laser cooling, atomic clock, photocell, nuclear medicine, and specialty glass.

Application

Rubidium can be used as a starting material to synthesize rubidium hydrazinidoborane (RbN2H3BH3).
It can also be used as a reducing agent to synthesize reduced polycyclic aromatic hydrocarbons.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A D Tranter et al.
Nature communications, 9(1), 4360-4360 (2018-10-21)
Machine learning based on artificial neural networks has emerged as an efficient means to develop empirical models of complex systems. Cold atomic ensembles have become commonplace in laboratories around the world, however, many-body interactions give rise to complex dynamics that
Zhifeng Chen et al.
Biosensors & bioelectronics, 41, 519-525 (2012-10-30)
A highly sensitive and simple label-free electrochemiluminescent (ECL) sensing strategy has been developed for assay of protein kinase A (PKA) activity and inhibition by taking advantage of zirconium cation (Zr(4+)) mediated signal transition and signal amplification of Ru(II) encapsulated phosphorylate-terminated
Hongmei Cao et al.
Biosensors & bioelectronics, 41, 911-915 (2012-11-06)
A novel melamine electrochemiluminescence (ECL) sensor was developed based on mesoporous SiO(2) nanospheres/Ru(bpy)(3)(2+)/Nafion modified electrodes. The homogeneous mesoporous silica nanospheres, synthesized using modified Stöber sol-gel process, were characterized by Field Emission Scanning Electron Microscopy (FE-SEM), Transmission Electron Microscopy (TEM) and
Heidi M Sampson et al.
Orphanet journal of rare diseases, 8, 11-11 (2013-01-16)
Many genetic diseases are due to defects in protein trafficking where the mutant protein is recognized by the quality control systems, retained in the endoplasmic reticulum (ER), and degraded by the proteasome. In many cases, the mutant protein retains function
Marco Bianchi et al.
ACS nano, 6(8), 7009-7015 (2012-07-31)
Rubidium adsorption on the surface of the topological insulator Bi(2)Se(3) is found to induce a strong downward band bending, leading to the appearance of a quantum-confined two-dimensional electron gas state (2DEG) in the conduction band. The 2DEG shows a strong

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service