Skip to Content
Merck
All Photos(2)

Key Documents

27497

Sigma-Aldrich

Tributyl citrate

greener alternative

≥97.0%

Synonym(s):

2-Hydroxy-1,2,3-propanetricarboxylic acid tributyl ester, Tri-n-butyl citrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H32O7
CAS Number:
Molecular Weight:
360.44
Beilstein:
1806072
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0%

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.445

density

1.043 g/mL at 20 °C (lit.)

functional group

ester
hydroxyl

greener alternative category

SMILES string

CCCCOC(=O)CC(O)(CC(=O)OCCCC)C(=O)OCCCC

InChI

1S/C18H32O7/c1-4-7-10-23-15(19)13-18(22,17(21)25-12-9-6-3)14-16(20)24-11-8-5-2/h22H,4-14H2,1-3H3

InChI key

ZFOZVQLOBQUTQQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tributyl citrate is a common solvent and plasticizer for polymers like PVC and its copolymers that are used in food wrapping films. Also, it is a common plasticizer employed in food contact materials, cosmetics, and food additives.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product can replace highly toxic diluents in membrane preparation and thus aligns with "Less Hazardous Chemical Syntheses". Click here for more information.

Application

Tributyl citrate can be used:
  • To incorporate in zein film to enhance its mechanical properties for industrial processing.
  • As a plasticizer to improve the ductile properties of poly(lactide) polymers.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improved mechanical property and water resistance of zein films by plasticization with tributyl citrate
Shi K, et al.
Journal of Agricultural and Food Chemistry, 60, 5988-5993 (2012)
Synthesis of tributyl citrate using acid ionic liquid as catalyst
Junming XU, et al.
Process Safety and Environmental Protection, 88, 28-30 (2010)
New development on plasticized poly (lactide): Chemical grafting of citrate on PLA by reactive extrusion
Hassouna F, et al.
European Polymer Journal, 48, 404-415 (2012)
Ke Shi et al.
Journal of agricultural and food chemistry, 60(23), 5988-5993 (2012-05-10)
Pure zein film is intrinsically rigid and brittle and lacks necessary mechanical properties for industrial processing. In addition, pure zein film is sensitive to high relative humidity, which limits its application in food packaging. To improve these properties, tributyl citrate
Eva Roblegg et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 79(3), 635-645 (2011-08-02)
The objective of this study was the development of retarded release pellets using vegetable calcium stearate (CaSt) as a thermoplastic excipient. The matrix carrier was hot melt extruded and pelletized with a hot-strand cutter in a one step continuous process.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service