Skip to Content
Merck
All Photos(1)

Key Documents

273619

Sigma-Aldrich

1′,3′-Dihydro-1′,3′,3′-trimethyl-6-nitrospiro[2H-1-benzopyran-2,2′-(2H)-indole]

98%

Synonym(s):

1′,3′,3′-trimethyl-6-nitrospiro[chromene-2,2′-indole], 1,3,3-Trimethylindolino-6′-nitrobenzopyrylospiran; 1′,3′,3′-Trimethyl-6-nitrospiro[chromene-2,2′-indoline]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18N2O3
CAS Number:
Molecular Weight:
322.36
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

solid

mp

179-180 °C (lit.)

λmax

242 nm in ethanol

SMILES string

CN1c2ccccc2C(C)(C)C13Oc4ccc(cc4C=C3)[N+]([O-])=O

InChI

1S/C19H18N2O3/c1-18(2)15-6-4-5-7-16(15)20(3)19(18)11-10-13-12-14(21(22)23)8-9-17(13)24-19/h4-12H,1-3H3

InChI key

PSXPTGAEJZYNFI-UHFFFAOYSA-N

General description

1′,3′-Dihydro-1′,3′,3′-trimethyl-6-nitrospiro(2H-1-benzopyran-2,2′-(2H)-indole)(NitroBIPS) is a photochromic biocompatible spiropyran molecule that has six nitro groups positioned at the benzopyran section. It has a colourless leuco spiropyran form(SP) and coloured trans-merocyanine form(MC) that exhibits absorption peaks in the ultraviolet and visible regions respectively. NitroBIPS possesses a higher quantum yield in comparison to other spiropyran molecules and dissolves in a variety of solvents.

Application

NitroBIPS is used as a photochromic dye which can be incorporated in poly(vinylalcohol)(PVA) by electro-spinning to form PVA-spiropyran nanofibres. It has photo-responsive behaviour that may be useful in optical data storage applications. It can be used to dope liquid crystal to develop a coloured scattering device that is dual responsive and allows the initiation of electrohydrodynamic instabilities(EHDI). Addition of NitroBIPS can modify the transparency from 100% to 16%, providing a shielding effect which can be used for smart window applications. It may also be used to enhance the adhesion property of polystyrene(PS) on the glass substrate by improving the shear strength by a factor of 7.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation, characterization, and application of optical switch probes.
Petchprayoon C and Marriott G
Current Protocols in Chemical Biology, 2(3), 153-169 (2010)
Direct structural determination of conformations of photoswitchable molecules by laser desorption-electron diffraction.
Andreas Gahlmann et al.
Angewandte Chemie (International ed. in English), 49(37), 6524-6527 (2010-08-18)
Xiang Xu et al.
Scandinavian journal of gastroenterology, 37(3), 330-337 (2002-03-28)
Mast cells have been implicated in chronic inflammatory conditions resulting in fibrosis, such as Crohn disease. However, a link between inflammation, fibrosis and mast cells has not been demonstrated in human or animal intestinal diseases. This work was undertaken to
K Poots et al.
Annals of biomedical engineering, 14(3), 203-218 (1986-01-01)
A nonintrusive method for the visualization of pulsatile flow velocity profiles is described. The method is based on the use of a photochromic dye that is added to the fluid being studied and a nitrogen laser which excites the dye
M J Jozwiakowski et al.
Pharmaceutical research, 4(5), 398-401 (1987-10-01)
The title compound (6-NO2-BIPS) is adsorptiochromic, becoming colored upon adsorption to a polar surface. Powders of 6-NO2-BIPS adsorbed to silica gel or silicic acid are suspended in glycerin, and the absorption spectrum of the adsorbate is recorded by conventional absorption

Articles

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service