Recommended Products
Quality Level
Assay
95%
form
liquid
refractive index
n20/D 1.49 (lit.)
bp
244 °C (lit.)
mp
31-32 °C (lit.)
density
0.86 g/mL at 25 °C (lit.)
SMILES string
Cc1cc(cc(c1)C(C)(C)C)C(C)(C)C
InChI
1S/C15H24/c1-11-8-12(14(2,3)4)10-13(9-11)15(5,6)7/h8-10H,1-7H3
InChI key
WIXDSJRJFDWTNY-UHFFFAOYSA-N
Application
3,5-Di-tert-butyltoluene was used in the synthesis of 3,5-di-tert-butyl(bromomethyl)benzene. It was also used in the synthesis of di-tert-butylbenzoic acid.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
THE SYNTHESIS AND SUBSTITUTION REACTIONS OF 3, 5-DI-tert-BUTYLPHENOL.
Canadian Journal of Chemistry, 41(7), 1653-1656 (1963)
A new synthetic route to donor-acceptor porphyrins.
Tetrahedron, 58(12), 2405-2413 (2002)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service