Skip to Content
Merck
All Photos(2)

Key Documents

268038

Sigma-Aldrich

Methyl oleate

technical grade, 70%

Synonym(s):

Methyl cis-9-octadecenoate, Oleic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)7CO2CH3
CAS Number:
Molecular Weight:
296.49
Beilstein:
1727037
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

vapor pressure

10 mmHg ( 205 °C)

Assay

70%

form

liquid

refractive index

n20/D 1.452 (lit.)

bp

218 °C/20 mmHg (lit.)

density

0.874 g/mL at 20 °C (lit.)

functional group

ester

SMILES string

CCCCCCCC\C=C/CCCCCCCC(=O)OC

InChI

1S/C19H36O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h10-11H,3-9,12-18H2,1-2H3/b11-10-

InChI key

QYDYPVFESGNLHU-KHPPLWFESA-N

Looking for similar products? Visit Product Comparison Guide

General description

The methyl oleate monolayers at the air-water interface undergoes ozonolysis that results in rapid loss of material through cleavage of the C[double bond]C bond and evaporation/dissolution of reaction products.

Application

Methyl oleate was used to study the catalytic potential of molybdenum oxide/bipyridinedicarboxylate hybrid material.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tatiana R Amarante et al.
Inorganic chemistry, 52(8), 4618-4628 (2013-03-28)
The reaction of MoO3, 2,2'-bipyridine-5,5-dicarboxylic acid (H2bpdc), water, and dimethylformamide in the mole ratio 1:1:1730:130 at 150 °C for 3 days in a rotating Teflon-lined digestion bomb leads to the isolation of the molybdenum oxide/bipyridinedicarboxylate hybrid material (DMA)[MoO3(Hbpdc)]·nH2O (1) (DMA
Hailey M Summers et al.
Bioresource technology, 196, 431-440 (2015-08-16)
The economic feasibility and environmental impact is investigated for the conversion of agricultural waste, delactosed whey permeate, through yeast fermentation to a renewable diesel via hydrothermal liquefaction. Process feasibility was demonstrated at laboratory-scale with data leveraged to validate systems models
Christian Pfrang et al.
Physical chemistry chemical physics : PCCP, 16(26), 13220-13228 (2014-05-30)
Ozonolysis of methyl oleate monolayers at the air-water interface results in surprisingly rapid loss of material through cleavage of the C=C bond and evaporation/dissolution of reaction products. We determine using neutron reflectometry a rate coefficient of (5.7 ± 0.9) ×
Annapurna Kumari et al.
Biotechnology for biofuels, 2(1), 1-1 (2009-01-16)
Transesterification of Jatropha oil was carried out in t-butanol solvent using immobilized lipase from Enterobacter aerogenes. The presence of t-butanol significantly reduced the negative effects caused by both methanol and glycerol. The effects of various reaction parameters on transesterification of
Ursula Biermann et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(26), 8201-8207 (2012-05-18)
A kinetic study of the dodecanethiol-catalyzed cis/trans isomerization of methyl oleate (cis-2) without added initiator was performed by focusing on the initiation of the radical chain reaction. The reaction orders of the rate of isomerization were 2 and 0.5 for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service