Skip to Content
Merck
All Photos(1)

Key Documents

264326

Sigma-Aldrich

2,4-Difluorobenzonitrile

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3CN
CAS Number:
Molecular Weight:
139.10
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

47-49 °C (lit.)

functional group

fluoro

SMILES string

Fc1ccc(C#N)c(F)c1

InChI

1S/C7H3F2N/c8-6-2-1-5(4-10)7(9)3-6/h1-3H

InChI key

LJFDXXUKKMEQKE-UHFFFAOYSA-N

General description

2,4-Difluorobenzonitrile undergoes polycondensation with Bisphenol A and matrix-assisted laser desorption/time-of-flight mass spectra revealed a quantitative formation of cyclic oligoethers and polyethers.

Application

2,4-Difluorobenzonitrile has been used in the synthesis of:
  • 2-methylsulfonyl-4-fluorobenzylamine
  • fluoro-3-amino-1,2-benzisoxazoles

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Macrocycles. XXVIII. Cyclic poly (benzonitrile ether) s derived from bisphenol A.
Kricheldorf HR, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 41(23), 3838-3846 (2003)
Practical Synthesis of 4-Fluoro-2-(methylthio) benzylamine and the Corresponding Sulfone and Sulfonamide.
Perlow DS, et al.
Synthetic Communications, 37(11), 1887-1897 (2007)
S D Lepore et al.
The Journal of organic chemistry, 65(10), 2924-2932 (2000-05-18)
Further exploration of the scope of our solid-phase method for the synthesis of 3-aminobenzisoxazoles (using the Kaiser oxime resin 1) is described. The effects of base, leaving group, and solvent on the nucleophilic aromatic substitution based resin-loading reaction are discussed.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service