Skip to Content
Merck
All Photos(2)

Key Documents

264237

Sigma-Aldrich

1,2-Dibromo-4,5-difluorobenzene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br2C6H2F2
CAS Number:
Molecular Weight:
271.88
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

mp

33-35 °C (lit.)

functional group

bromo
fluoro

SMILES string

Fc1cc(Br)c(Br)cc1F

InChI

1S/C6H2Br2F2/c7-3-1-5(9)6(10)2-4(3)8/h1-2H

InChI key

JTEZQWOKRHOKDG-UHFFFAOYSA-N

Application

1,2-Dibromo-4,5-difluorobenzene has been used in the preparation of:
  • 2,2′-dibromo-4,4′,5,5′-tetrafluorobiphenyl
  • 2-(2,3-dibromo-5,6-difluorophenyl)propan-2-ol

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

111.2 °F - closed cup

Flash Point(C)

44 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthesis of biphenylenes and tetraphenylenes using copper-catalyzed coupling of arylzinc intermediates.
Kabir SMH, et al.
Journal of the Chemical Society. Perkin Transactions 1, 2, 159-165 (2001)
The lithiation of fluorinated benzenes and its dependence on solvent and temperature.
Coe PL, et al.
Journal of the Chemical Society. Perkin Transactions 1, 21, 2729-2737 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service