263389
3,5-Bis(trifluoromethyl)benzylamine
technical grade, 80%
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Recommended Products
grade
technical grade
Assay
80%
form
solid
mp
50-55 °C (lit.)
functional group
amine
fluoro
SMILES string
NCc1cc(cc(c1)C(F)(F)F)C(F)(F)F
InChI
1S/C9H7F6N/c10-8(11,12)6-1-5(4-16)2-7(3-6)9(13,14)15/h1-3H,4,16H2
InChI key
DHVHORCFFOSRBP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
3,5-Bis(trifluoromethyl)benzylamine has been used in the preparation of phenyl glycine derivatives.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
172.4 °F - closed cup
Flash Point(C)
78 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Bioorganic & medicinal chemistry letters, 16(14), 3735-3739 (2006-05-16)
Systematic modification of a screening lead yielded a class of potent glycinamide based CCR2 antagonists. The best compound (55, (2S)-N-[3,5-bis(trifluoromethyl)benzyl]-2-{[2-(1-piperidinyl)ethyl]amino}-2-(3-thienyl)acetamide) displayed good binding affinity (IC50=30 and 39 nM) toward human monocytes and CHO cell expressing human CCR2b, respectively. Functionally, it
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service