Skip to Content
Merck
All Photos(1)

Key Documents

262536

Sigma-Aldrich

trans-3-Nonen-2-one

95%

Synonym(s):

(3E)-3-Nonen-2-one, (3E)-Non-3-en-2-one, (E)-3-Nonen-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH=CHCOCH3
CAS Number:
Molecular Weight:
140.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

refractive index

n20/D 1.449 (lit.)

bp

85 °C/12 mmHg (lit.)

density

0.848 g/mL at 25 °C (lit.)

functional group

ketone

SMILES string

[H]\C(CCCCC)=C(\[H])C(C)=O

InChI

1S/C9H16O/c1-3-4-5-6-7-8-9(2)10/h7-8H,3-6H2,1-2H3/b8-7+

InChI key

HDKLIZDXVUCLHQ-BQYQJAHWSA-N

Application

trans-3-Nonen-2-one has been used:
  • as substrate to investigate steady-state kinetics of NADPH oxidation
  • in the preparation of azido quinolone

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryan A Dick et al.
The Journal of biological chemistry, 279(17), 17269-17277 (2004-02-18)
NADPH-dependent alkenal/one oxidoreductase (AOR) from the rat is a phase 2/antioxidative enzyme that is known to catalyze the reduction of the carbon-carbon double bond of alpha,beta-unsaturated aldehydes and ketones. It is also known for its leukotriene B(4) 12-hydroxydehydrogenase activity. In
Hajoong Lee et al.
The Journal of organic chemistry, 75(5), 1756-1759 (2010-02-10)
We describe the assembly of a 960-member library of tricyclic 2,3-dihydro-4-quinolones using a combination of solution-phase high-throughput organic synthesis and parallel chromatographic purification. The library was produced with high efficiency and complete chemo- and diastereoselectivity by diversification of an azide-bearing
Emiliano Ventura et al.
Journal of chromatography. A, 1600, 183-196 (2019-05-06)
A semi-quantitative method was developed to monitor the misuse of 15 SARM compounds belonging to nine different families, in urine matrices from a range of species (equine, canine, human, bovine and murine). SARM residues were extracted from urine (200 μL) with
Emiliano Ventura et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 37(8), 1253-1263 (2020-05-14)
Selective androgen receptor modulators (SARMs) are a class of new emerging "designer" steroid compounds gaining popularity over more well established anabolic-androgenic steroids (AAS) amongst both non-professional and elite athletes. Moreover, due to their anabolic activity, SARM compounds may also potentially

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service