Skip to Content
Merck
All Photos(1)

Key Documents

261939

Sigma-Aldrich

1,2-Bis(dimethylphosphino)ethane

97%

Synonym(s):

DMPE, Dimethyl-2-(dimethylphosphino)ethylphosphine, Ethylenebis(dimethylphosphine)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2PCH2CH2P(CH3)2
CAS Number:
Molecular Weight:
150.14
Beilstein:
1732994
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

refractive index

n20/D 1.507 (lit.)

bp

180 °C (lit.)

density

0.9 g/mL at 25 °C (lit.)

functional group

phosphine

SMILES string

CP(C)CCP(C)C

InChI

1S/C6H16P2/c1-7(2)5-6-8(3)4/h5-6H2,1-4H3

InChI key

ZKWQSBFSGZJNFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

1,2-Bis(dimethylphosphino)ethane (dmpe) is a polydentate phosphine ligand. Synthesis of 1,2-bis(dimethylphosphino)ethane has been reported. It reacts with ZrCl2 to afford ZrCl4(dmpe)2 complexes.

Application

1,2-Bis(dimethylphosphino)ethane (dmpe) may be employed as ligand in the preparation of cis-arylmethyl(dmpe) nickel(II) complexes.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A convenient synthesis of 1, 2-bis (dichlorophosphino) ethane, 1, 2-bis (dimethylphosphino) ethane and 1,2-bis (diethylphosphino) ethane.
Burt RJ, et al.
Journal of Organometallic Chemistry, 182(2), 203-206 (1979)
Phosphine-induced reductive elimination from cis-arylmethylnickel (II) complexes having a 1,2-bis(dimethylphosphino) ethane ligand
Komiya S, et al.
Organometallics, 2(10), 1466-1468 (1983)
Beatty RP, et al.
Inorganic Chemistry, 18(11), 3139-3145 (1979)
M Suwalsky et al.
Biochimica et biophysica acta, 1848(11 Pt A), 2829-2838 (2015-08-25)
Usnic acid (UA) has been associated with chronic diseases through its antioxidant action. Its main target is the cell membrane; however, its effect on that of human erythrocytes has been scarcely investigated. To gain insight into the molecular mechanisms of
Mario Suwalsky et al.
The Journal of membrane biology, 248(4), 683-693 (2015-03-01)
Despite the extended use and well-documented information, there are insufficient reports concerning the effects of propranolol on the structure and functions of cell membranes, particularly those of human erythrocytes. Aimed to better understand the molecular mechanisms of its interactions with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service