Skip to Content
Merck
All Photos(2)

Key Documents

261416

Sigma-Aldrich

4-Chloro-2-isopropyl-5-methylphenol

99%

Synonym(s):

Chlorothymol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHC6H2(Cl)(CH3)OH
CAS Number:
Molecular Weight:
184.66
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

60-62 °C (lit.)

solubility

ethanol: soluble 10%, clear to very slightly hazy, yellow
water: insoluble

functional group

chloro

SMILES string

CC(C)c1cc(Cl)c(C)cc1O

InChI

1S/C10H13ClO/c1-6(2)8-5-9(11)7(3)4-10(8)12/h4-6,12H,1-3H3

InChI key

KFZXVMNBUMVKLN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Photocatalytic degradation of 4-chloro-2-isopropyl-5-methylphenol has been investigated in water and in simulated soil washing wastes containing Brij 35 in the presence of TiO2 dispersions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Davezza et al.
Environmental science and pollution research international, 20(5), 3224-3231 (2012-10-16)
The photocatalytic degradations of 4-chlorophenol (CP), 4-chloro-2-methylphenol (CMP), 4-chloro-3,5-dimethylphenol (CDMP) and 4-chloro-2-isopropyl-5-methylphenol (CIMP) were investigated in water and in simulated soil washing wastes containing Brij 35 (polyoxyethylene(23)dodecyl ether) in the presence of TiO2 dispersions. A neat inhibition of substrate decomposition

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service