All Photos(1)
About This Item
Linear Formula:
ClCH2CONCO
CAS Number:
Molecular Weight:
119.51
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
≥95%
form
liquid
refractive index
n20/D 1.463 (lit.)
bp
50-55 °C/20 mmHg (lit.)
density
1.403 g/mL at 25 °C (lit.)
functional group
amine
chloro
isocyanate
storage temp.
−20°C
SMILES string
ClCC(=O)N=C=O
InChI
1S/C3H2ClNO2/c4-1-3(7)5-2-6/h1H2
InChI key
MOVMEFHWBOWMFU-UHFFFAOYSA-N
Related Categories
Application
Chloroacetyl isocyanate has been used in the synthesis of:
- fumagillin analogs
- 2-chloro-N-(4-oxocyclopent-2-enyl)-acetamide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Resp. Sens. 1 - Skin Corr. 1B
Supplementary Hazards
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
143.6 °F - closed cup
Flash Point(C)
62 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Synthesis of 4-azacyclopent-2-enones and 5, 5-dialkyl-4-azacyclopent-2-enones.
Dauvergne J, et al.
Tetrahedron, 60(11), 2559-2567 (2004)
Ralph Mazitschek et al.
Organic & biomolecular chemistry, 3(11), 2150-2154 (2005-05-27)
A promising approach among the numerous efforts to cure cancer is the interruption of the tumour-induced formation of new blood vessels (angiogenesis). By suppressing angiogenesis with drugs, the tumour can neither grow to a life threatening size, nor metastasize. The
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service