Skip to Content
Merck
All Photos(1)

Key Documents

258903

Sigma-Aldrich

3,4-Dichloro-1,2,5-thiadiazole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2Cl2N2S
CAS Number:
Molecular Weight:
155.01
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.561 (lit.)

bp

158 °C (lit.)

density

1.648 g/mL at 25 °C (lit.)

SMILES string

Clc1nsnc1Cl

InChI

1S/C2Cl2N2S/c3-1-2(4)6-7-5-1

InChI key

YNZQOVYRCBAMEU-UHFFFAOYSA-N

Application

3,4-Dichloro-1,2,5-thiadiazole has been employed:
  • as guest molecule to investigate solid-state 13C NMR spectra of inclusion compounds of 2,6-dimethyl-bicyclo[3.3.1]nonane-exo-2-exo-6-diol with small organic molecules
  • in preparation of [1,2,5]thiadiazol-3-yl-piperazine compounds

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

D Gizachew et al.
Solid state nuclear magnetic resonance, 3(2), 67-78 (1994-04-01)
Solid-state 13C nuclear magnetic resonance (NMR) spectra of a number of inclusion compounds of 2,6-dimethyl-bicyclo[3.3.1]nonane-exo-2-exo-6-diol (host) with small organic small molecules (guests) have been studied. With 3,4-dichloro-1,2,5-thiadiazole and tetrachloroethylene as guests, line splittings of the host resonances were observed due
D Srinivas et al.
Rapid communications in mass spectrometry : RCM, 31(1), 121-128 (2016-10-28)
1,2,5-Thiadiazoles are an important class of compounds mostly used in synthetic chemistry, and as herbicides, insecticides, drugs, organic conductors, etc. Recently, they have been used as a source for the generation and study of nitrile N-sulfides, RCNS, and its isomers.
A L Sabb et al.
Bioorganic & medicinal chemistry letters, 11(8), 1069-1071 (2001-05-01)
Amino acid derivatives of 1,2,5-thiadiazol-3-yl-piperazine related to (+)-WAY-100135 and WAY-100635 are potent 5-HT1A receptor agonists and antagonists, which have selective affinity for 5-HT1A receptors versus alpha1 and dopamine (D2, D3, and D4) receptors.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service