Skip to Content
Merck
All Photos(1)

Key Documents

256560

Sigma-Aldrich

1-Pentyne

99%

Synonym(s):

Propylacetylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2C≡CH
CAS Number:
Molecular Weight:
68.12
Beilstein:
1697133
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

6.8 psi ( 20 °C)

Assay

99%

form

liquid

refractive index

n20/D 1.385 (lit.)

bp

40 °C (lit.)

mp

−106-−105 °C (lit.)

density

0.691 g/mL at 25 °C (lit.)

SMILES string

CCCC#C

InChI

1S/C5H8/c1-3-5-4-2/h1H,4-5H2,2H3

InChI key

IBXNCJKFFQIKKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Selective and non-selective hydrogenation of 1-pentyne catalyzed by silica-supported palladium has been studied by in situ X-ray absorption spectroscopy.

Application

1-Pentyne has been used in preparation of:
  • lithium acetylides, required for asymmetric synthesis of α,α-dibranched propargyl sulfinamides
  • 7-hydroxy-10-methoxy-3H-naphtho[2.1-b]pyrans

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Min Wei Tew et al.
Physical chemistry chemical physics : PCCP, 14(16), 5761-5768 (2012-03-17)
The catalytically active phase of silica-supported palladium catalysts in the selective and non-selective hydrogenation of 1-pentyne was determined using in situ X-ray absorption spectroscopy at the Pd K and L(3) edges. Upon exposure to alkyne, a palladium carbide-like phase rapidly
Manish Rawat et al.
Journal of the American Chemical Society, 128(34), 11044-11053 (2006-08-24)
The carbene complex 5-(2,2-dimethyl-2H-chromene)methoxylmethylene chromium pentacarbonyl will undergo a benzannulation reaction with phenylacetylene, 1-pentyne, 3-hexyne, and trimethylsilylacetylene to give 7-hydroxy-10-methoxy-3H-naphtho[2.1-b]pyrans as the primary product. These compounds are difficult to obtain pure due to their sensitivity to air. If the benzannulation
Andrew W Patterson et al.
The Journal of organic chemistry, 71(18), 7110-7112 (2006-08-26)
Addition of lithium acetylides prepared from 1-pentyne, phenylacetylene, and trimethylsilylacetylene to diverse N-tert-butanesulfinyl ketimines affords a range of alpha,alpha-dibranched propargyl sulfinamides in generally good yields (up to 87%) and with high diastereoselectivities (up to >99:1). Acidic cleavage of the tert-butanesulfinyl
Quan-De Wang et al.
International journal of molecular sciences, 20(13) (2019-07-03)
Hydrogen atom abstraction from propargyl C-H sites of alkynes plays a critical role in determining the reactivity of alkyne molecules and understanding the formation of soot precursors. This work reports a systematic theoretical study on the reaction mechanisms and rate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service