Skip to Content
Merck
All Photos(1)

Key Documents

251739

Sigma-Aldrich

3-Allyloxy-1,2-propanediol

99%

Synonym(s):

Glycerol α-monoallyl ether, Glycerol 1-allyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(OH)CH2OCH2CH=CH2
CAS Number:
Molecular Weight:
132.16
Beilstein:
1701144
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

liquid

refractive index

n20/D 1.462 (lit.)

bp

142 °C/28 mmHg (lit.)

density

1.068 g/mL at 25 °C (lit.)

SMILES string

OCC(O)COCC=C

InChI

1S/C6H12O3/c1-2-3-9-5-6(8)4-7/h2,6-8H,1,3-5H2

InChI key

PAKCOSURAUIXFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Allyloxy-1,2-propanediol(AP) is a multifunctional monomer which contains both allyl and hydroxyl functionalities. The allyl group allows for easy incorporation into polymer chains through various polymerization techniques, including free radical polymerization and thiol-ene reactions. The hydroxyl groups contribute to its solubility in water and compatibility with hydrophilic polymers, enhancing its utility in biomedical applications. It also exhibits high thermal stability.

Application

3-Allyloxy-1,2-propanediol can be used:
  • As a functional additive to fabricate spin-coating-free perovskite photovoltaic devices. The hydroxyl groups of the AP interact with the perovskite, resulting in the creation of a high-quality slot-die coated film. This suppresses surface defects in the perovskite, enhancing the performance and stability.
  • As a monomer to synthesize modified polymer grafted magnetic nanoparticles via radical co-polymerization method. This nano-adsorbent with high chemical stability can be used in targeted drug delivery systems. It enables the targeted adsorption and extraction of specific molecules.
  • As a precursor to prepare hyper-branched solid polymer electrolytes for long-life rechargeable Li-ion batteries.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nathan Z Dreger et al.
Biomaterials, 221, 119399-119399 (2019-08-20)
Hernia repair outcomes have improved with more robust material options for surgeons and optimized surgical techniques. However, ventral hernia repairs remain challenging with an inherent risk of post-surgical adhesions in the peritoneal space which can occur regardless of interventional material
Chunhui Ruan et al.
ACS applied materials & interfaces, 10(12), 10398-10408 (2018-03-03)
An ideal gene-carrying vector is supposed to exhibit outstanding gene-condensing capability with positively charged macromolecules to protect the carried gene during in vivo circulation and a rapid dissociation upon microenvironmental stimuli at the aimed sites to release the escorted gene.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service