Skip to Content
Merck
All Photos(1)

Key Documents

249696

Sigma-Aldrich

2-Amino-4-thiazoleacetic acid

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6N2O2S
CAS Number:
Molecular Weight:
158.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

powder

mp

130 °C (dec.) (lit.)

functional group

carboxylic acid

storage temp.

−20°C

SMILES string

Nc1nc(CC(O)=O)cs1

InChI

1S/C5H6N2O2S/c6-5-7-3(2-10-5)1-4(8)9/h2H,1H2,(H2,6,7)(H,8,9)

InChI key

DYCLHZPOADTVKK-UHFFFAOYSA-N

Application

2-Amino-4-thiazoleacetic acid has been used in:
  • preparation of poly(2-amino-4-thiazoleacetic acid)/multiwalled carbon nanotubes modified glassy carbon electrodes, for the voltammetric determination of copper ions
  • electrochemical synthesis of novel thiozole-based copolymer with 2-aminothiazole
  • preparation of aquabis(2-amino-1,3-thiazole-4-acetato-κ2O,N3)nickel (II)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiu-Fen He et al.
Acta crystallographica. Section E, Structure reports online, 65(Pt 6), m666-m666 (2009-01-01)
In the crystal structure of the title compound, [Ni(C(5)H(5)N(2)O(2)S)(2)(H(2)O)], the Ni(II) cation is located on a twofold rotation axis and chelated by two 2-amino-1,3-thia-zole-4-acetate (ata) anions in the basal coordination plane; a water mol-ecule located on the same twofold rotation
Poly (2-amino-4-thiazoleacetic acid)/multiwalled carbon nanotubes modified glassy carbon electrodes for the electrochemical detection of copper (II).
Zhao H, et al.
Electrochimica Acta, 55(7), 256-263 (2010)
Electrochemical synthesis and characterization of a novel thiazole-based copolymer and its application in biosensor.
Xue Y, et al.
Electrochimica Acta, 59, 256-263 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service