Skip to Content
Merck
All Photos(1)

Key Documents

247154

Sigma-Aldrich

3,4-Dibromothiophene

99%

Synonym(s):

3,4-Dibromothiophene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H2Br2S
CAS Number:
Molecular Weight:
241.93
Beilstein:
107642
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.640 (lit.)

bp

221-222 °C (lit.)

mp

4-5 °C (lit.)

density

2.188 g/mL at 25 °C (lit.)

functional group

bromo

SMILES string

Brc1cscc1Br

InChI

1S/C4H2Br2S/c5-3-1-7-2-4(3)6/h1-2H

InChI key

VGKLVWTVCUDISO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Double photoionization spectra of 3,4-dibromothiophene has been investigated by coincidence spectroscopy technique based on electron time-of-flight measurement.

Application

3,4-Dibromothiophene was used in the preparation of thieno[3,4-b]thiophene. It was aslo used as starting material in the synthesis of alkyl substituted, fused thiophenes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An Alternative Synthysis of Thieno [3, 4-b] Thiophene.
Brandsma L and Verkruijsse HD.
Synthetic Communications, 20(15), 2275-2277 (1990)
P Linusson et al.
The Journal of chemical physics, 129(23), 234303-234303 (2008-12-24)
We report the double photoionization spectra of thiophene, 3-bromothiophene, and 3,4-dibromothiophene using a coincidence spectroscopy technique based on electron time-of-flight measurements. Spectra have been recorded between the onset and 40.814 eV using He IIalpha radiation. The He I photoelectron spectrum
Mingqian He et al.
The Journal of organic chemistry, 72(2), 442-451 (2007-01-16)
We have established a series of synthetic methods to synthesize alkyl-substituted fused thiophenes with degrees of fusion from two to seven rings. These fused thiophene ring compounds have very good solubility in common organic solvents, making possible the solution processing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service