Skip to Content
Merck

246549

Sigma-Aldrich

Diethylmalonic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2C(COOH)2
CAS Number:
Molecular Weight:
160.17
Beilstein:
1768554
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Pricing and availability is not currently available.

Assay

98%

form

powder

mp

129-131 °C (lit.)

SMILES string

CCC(CC)(C(O)=O)C(O)=O

InChI

1S/C7H12O4/c1-3-7(4-2,5(8)9)6(10)11/h3-4H2,1-2H3,(H,8,9)(H,10,11)

InChI key

LTMRRSWNXVJMBA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O Jáuregui et al.
Electrophoresis, 21(3), 611-618 (2000-03-22)
Optimum conditions for the separation of positional isomers of chlorophenols by capillary zone electrophoresis (CZE) were established. The behavior of five volatile electrolytes (L-cysteic acid, 3-amino-1-propanesulfonic acid, aminomethanesulfonic acid, diethylmalonic acid, and ammonium acetate) was compared. The best performance based
C Chevanet et al.
Canadian journal of microbiology, 32(3), 254-258 (1986-03-01)
Bacillomycin L is produced by Bacillus subtilis NCIB 8872 in the stationary phase; it is excreted into the culture medium, without prior accumulation in the bacterial cells. The production of bacillomycin L is largely dependent on the composition of the
W G Reifenrath et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 4(2 Pt 2), S224-S230 (1984-04-01)
The human skin grafted athymic nude mouse, pig skin grafted athymic nude mouse, hairless dog, and weanling Yorkshire pig were evaluated as models for predicting skin penetration in man. Nine radiolabeled compounds previously tested on man were applied topically (4
Roger Peró-Gascón et al.
Journal of chromatography. A, 1508, 148-157 (2017-06-18)
In this paper we describe a strategy to estimate by CE the acidity constants (pK
W G Reifenrath et al.
Journal of pharmaceutical sciences, 73(10), 1388-1392 (1984-10-01)
A shower decontamination bench model has been used to assess quantitatively the importance of several variables (water pressure and temperature, surfactant concentration in the decontamination fluid, nozzle type, and shower time) on decontamination of nontoxic chemical warfare-agent simulants diethyl malonate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service