About This Item
Recommended Products
Quality Level
Assay
99%
form
solid
mp
87-90 °C (lit.)
solubility
ethanol: 50 mg/mL, clear, colorless to faintly yellow
functional group
chloro
SMILES string
Clc1ccc2cc[nH]c2c1
InChI
1S/C8H6ClN/c9-7-2-1-6-3-4-10-8(6)5-7/h1-5,10H
InChI key
YTYIMDRWPTUAHP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service