Skip to Content
Merck
All Photos(1)

Key Documents

245976

Sigma-Aldrich

2,4,6-Tri-tert-butylpyridine

99%

Synonym(s):

2,4,6-Tris(1,1-dimethylethyl)pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H29N
CAS Number:
Molecular Weight:
247.42
Beilstein:
1620027
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

bp

115-120 °C/20 mmHg (lit.)

mp

67-71 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)(C)c1cc(nc(c1)C(C)(C)C)C(C)(C)C

InChI

1S/C17H29N/c1-15(2,3)12-10-13(16(4,5)6)18-14(11-12)17(7,8)9/h10-11H,1-9H3

InChI key

IDOFMGSESVXXQI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,4,6-Tri-tert-butylpyridine was used in the synthesis of indole triflones.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Naming of a child].
Y Endo
Josanpu zasshi = The Japanese journal for midwife, 32(1), 55-55 (1978-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service