Skip to Content
Merck
All Photos(3)

Key Documents

245585

Sigma-Aldrich

Sodium bis(trimethylsilyl)amide solution

1.0 M in THF

Synonym(s):

Hexamethyldisilazane sodium salt solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3Si]2NNa
CAS Number:
Molecular Weight:
183.37
Beilstein:
3629917
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

concentration

1.0 M in THF

density

0.904 g/mL at 25 °C

SMILES string

C[Si](C)(C)N([Na])[Si](C)(C)C

InChI

1S/C6H18NSi2.Na/c1-8(2,3)7-9(4,5)6;/h1-6H3;/q-1;+1

InChI key

WRIKHQLVHPKCJU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium bis(trimethylsilyl)amide solution (NaHMDS) is widely used as a strong base in organic synthesis for deprotonation reactions and base-catalyzed reactions. It is also involved in the generation of enolates, Wittig reagents and carbenes.

Application

NaHMDS can be used as a base:
  • In the synthesis of N-boc-1-amino-1,3-diene derivatives from N-boc-1-amino-4-methoxy-2-alkenes via 1,4-elimination reactions.
  • To deprotonate alkyl or aryl ketones to generate enolate derivatives by hydroxylation reaction.
  • In the enantioselective synthesis of 1,2-amino alcohols by the reaction between various nitroalkanes and aldehydes via nitroaldol reaction promoted by bimetallic heterogeneous catalyst.

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereoselective preparation of (1Z)-and (1E)-N-Boc-1-amino-1, 3-dienes by stereospecific base-promoted 1, 4-elimination
Tayama E and Toma Y
Tetrahedron, 71(4), 554-559 (2015)
Synthetic studies on (1S)-1-(6, 7-dimethoxy-2-naphthyl)-1-(1H-imidazol-4-yl)-2-methylpropan-1-ol as a selective C17, 20-lyase inhibitor
Matsunaga N, et al.
Tetrahedron Asymmetry, 15(13), 2021-2028 (2004)
Lithium and potassium bis (trimethylsilyl) amide: Utilizing non-nucleophilic bases as nitrogen sources
Bruning J, et al.
Tetrahedron Letters, 38(18), 3187-3188 (1997)
Macromolecules, 39, 5347-5347 (2006)
Elsa Vennat et al.
Dental materials : official publication of the Academy of Dental Materials, 25(6), 729-735 (2009-01-29)
The objectives of this study were to assess demineralized dentin porosity and quantify the different porous features distribution within the material using mercury intrusion porosimetry (MIP) technique. We compared hexamethyldisilazane (HMDS) drying and lyophilization (LYO) (freeze-drying) in sample preparation. Fifty-six

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service