244546
Azidomethyl phenyl sulfide
95%
Synonym(s):
Phenylthiomethyl azide
About This Item
Recommended Products
Assay
95%
reaction suitability
reaction type: click chemistry
refractive index
n20/D 1.5904 (lit.)
bp
104-105 °C/5 mmHg (lit.)
density
1.168 g/mL at 25 °C (lit.)
functional group
azide
thioether
storage temp.
2-8°C
SMILES string
[N-]=[N+]=NCSc1ccccc1
InChI
1S/C7H7N3S/c8-10-9-6-11-7-4-2-1-3-5-7/h1-5H,6H2
InChI key
KIQGRMGPIMCXSC-UHFFFAOYSA-N
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
226.4 °F - closed cup
Flash Point(C)
108 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service