243647
Molybdenum disilicide
powder, ≥99% trace metals basis
Synonym(s):
Molybdenum silicide
About This Item
Recommended Products
Quality Level
Assay
≥99% trace metals basis
form
powder
mp
1870-2030 °C (lit.)
density
6.31 g/mL at 25 °C (lit.)
SMILES string
[Mo++].[Si-]#[Si-]
InChI
1S/Mo.Si2/c;1-2/q+2;-2
InChI key
OVVBPJHTRZEJHC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
- Single-Crystal MoO3 Micrometer and Millimeter Belts Prepared from Discarded Molybdenum Disilicide Heating Elements.: This study explores the utilization of discarded molybdenum disilicide heating elements to synthesize single-crystal MoO3 belts. These materials exhibit high thermal stability and potential applications in high-temperature oxidation resistance and energy storage devices (Hou et al., 2018).
- Highly Concentrated Si1-xCx Alloy with an Ordered Superstructure.: The research investigates Si-C alloys with ordered superstructures, highlighting their potential as high-temperature semiconductors. This material′s high thermal resistance and electronic properties make it suitable for advanced semiconductor applications (Qin et al., 2003).
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Mechanical alloying is a “brute force” method of affecting alloying and chemical reactions. The mixture of reactant powders and several balls are placed in the milling jar of a high-energy ball mill, for example, a shaker mill or a planetary mill.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service