Skip to Content
Merck
All Photos(2)

Key Documents

238325

Sigma-Aldrich

Allyl iodide

98%

Synonym(s):

3-Iodo-1-propene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2I
CAS Number:
Molecular Weight:
167.98
Beilstein:
1697594
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

contains

copper as stabilizer

refractive index

n20/D 1.554 (lit.)

bp

102-103 °C (lit.)

solubility

alcohol: miscible(lit.)
chloroform: miscible(lit.)
diethyl ether: miscible(lit.)
water: insoluble (practically)(lit.)

density

1.837 g/mL at 25 °C (lit.)

functional group

alkyl halide

storage temp.

2-8°C

SMILES string

ICC=C

InChI

1S/C3H5I/c1-2-3-4/h2H,1,3H2

InChI key

HFEHLDPGIKPNKL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The interaction of allyl iodide with MoO3, Bi2O3 and molybdates of Bi, Co and Mg has been studied in the temperature range of 270-480°C.

Application

Allyl iodide has been employed as:
  • precursor for generation of allyl radicals to study their recombination in a single pulse shock tube with gas chromatographic measurements
  • reagent used with allylindium sesquiiodide in the cis-double allylation of cyclopropenes

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

60.8 °F - closed cup

Flash Point(C)

16 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interaction of allyl iodide with molybdate catalysts for the selective oxidation of hydrocarbons.
Grzybowska B, et al.
J. Catal., 49(2), 150-163 (1977)
Aleksandr Fridlyand et al.
The journal of physical chemistry. A, 117(23), 4762-4776 (2013-05-18)
The recombination and disproportionation of allyl radicals has been studied in a single pulse shock tube with gas chromatographic measurements at 1-10 bar, 650-1300 K, and 1.4-2 ms reaction times. 1,5-Hexadiene and allyl iodide were used as precursors. Simulation of
Xin Ma et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(25), 6355-6361 (2019-02-28)
Unexpectedly, the 5-dehydroquinoline radical cation was formed in the gas phase from the 5-iodo-8-nitroquinolinium cation upon ion-trap collision-activated dissociation. This reaction involves the cleavage of a nitro group to generate an intermediate monoradical, namely, the 8-dehydro-5-iodoquinolinium cation, followed by rearrangement
Tsunehisa Hirashita et al.
Organic & biomolecular chemistry, 5(13), 2154-2158 (2007-06-22)
The successive double allylation of cyclopropenes with allylindium sesquiiodide and allyl iodide proceeded with a cis-addition mode in the presence of other organometallics (e.g. Grignard reagent, cuprate, Et(2)Zn and Et(3)Al), giving the corresponding cis-diallylcyclopropanes in high yields.
Michael Lesslie et al.
European journal of mass spectrometry (Chichester, England), 21(3), 589-597 (2015-08-27)
Radical migration, both intramolecular and intermolecular, from the tyrosine phenoxyl radical Tyr(O(∙)) to the cysteine radical Cys(S(∙)) in model peptide systems was observed in the gas phase. Ion-molecule reactions (IMRs) between the radical cation of homotyrosine and propyl thiol resulted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service