Skip to Content
Merck
All Photos(2)

Key Documents

238155

Sigma-Aldrich

2-Bromoethyl methyl ether

Synonym(s):

1-Bromo-2-methoxyethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2CH2OCH3
CAS Number:
Molecular Weight:
138.99
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

refractive index

n20/D 1.447 (lit.)

bp

40-41 °C/66 mmHg (lit.)

solubility

water: soluble 14.4 g/L at 25 °C

density

1.479 g/mL at 25 °C (lit.)

functional group

bromo
ether

storage temp.

2-8°C

SMILES string

COCCBr

InChI

1S/C3H7BrO/c1-5-3-2-4/h2-3H2,1H3

InChI key

YZUPZGFPHUVJKC-UHFFFAOYSA-N

General description

Kinetics of potassium iodide exchange reaction with 2-bromoethyl methyl ether in acetone at 15 and 25°C has been investigated.

Application

2-Bromoethyl methyl ether has been used as starting reagent in the synthesis of 1-bromo-4-(2-methoxyethoxy)benzene and 1-bromo-4-[(2-methoxyethoxy)methyl]benzene.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

82.4 °F - closed cup

Flash Point(C)

28 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel water-soluble organosilane compounds as a radical reducing agent in aqueous media.
Yamazaki O, et al.
Bulletin of the Chemical Society of Japan, 70, 2519-2523 (1997)
Alex R Neale et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 18(15), 2040-2057 (2017-05-19)
A series of hydrophobic room temperature ionic liquids (ILs) based on ethereal functionalised pyrrolidinium, piperidinium and azepanium cations bearing the bis[(trifluoromethyl)sulfonyl]imide, [TFSI]
Ulderico Ulissi et al.
ChemSusChem, 11(1), 229-236 (2017-09-30)
The room-temperature molten salt mixture of N,N-diethyl-N-(2-methoxyethyl)-N-methylammonium bis(trifluoromethanesulfonyl) imide ([DEME][TFSI]) and lithium bis(trifluoromethanesulfonyl)imide (LiTFSI) salt is herein reported as electrolyte for application in Li-O
The Iodide Ion Exchange Reaction with 2-Haloethyl Alkyl Ethers and n-Butyl Bromide1.
Tutwiler FB and McKee RL.
Journal of the American Chemical Society, 76(24), 6342-6344 (1954)
Forrest S Gittleson et al.
Physical chemistry chemical physics : PCCP, 21(31), 17176-17189 (2019-07-26)
Ionic liquids are a unique class of materials with several potential applications in electrochemical energy storage. When used in electrolytes, these highly coordinating solvents can influence device performance through their high viscosities and strong solvation behaviors. In this work, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service