Skip to Content
Merck
All Photos(1)

Key Documents

237876

Sigma-Aldrich

Trimethyl orthoacetate

99%

Synonym(s):

1,1,1-Trimethoxyethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C(OCH3)3
CAS Number:
Molecular Weight:
120.15
Beilstein:
1098338
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.388 (lit.)

bp

107-109 °C (lit.)

density

0.944 g/mL at 25 °C (lit.)

functional group

ether

SMILES string

COC(C)(OC)OC

InChI

1S/C5H12O3/c1-5(6-2,7-3)8-4/h1-4H3

InChI key

HDPNBNXLBDFELL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Kinetics and mechanism of gas-phase elimination of trimethyl orthoacetate has been examined over the temperature range of 310-369°C and pressure range of 50-130Torr.

Trimethyl orthoacetate is used to synthesize acrylonitrile intermediates via reaction with alcohols followed by Knoevenagel condensation.

Application

Trimethyl orthoacetate has been used in the preparation of 2-amino-9-(3-acyloxymethyl-4-alkoxycarbonyloxybut-1-yl)purines and 2-amino-9-(3-alkoxycarbonyl-oxymethyl-4-alkoxycarbonyloxybut -1-yl)purines.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Sens. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transesterification of trimethyl orthoacetate: an efficient protocol for the synthesis of 4-alkoxy-2-aminothiophene-3-carbonitriles
Rao KV, et al.
Tetrahedron Letters, 54, 1274-1278 (2013)
Takahide Murakami et al.
Acta biomaterialia, 84, 257-267 (2018-12-12)
Postoperative adhesion is a relevant clinical problem that causes a variety of clinical complications after abdominal surgery. The objective of this study is to develop a liquid-type antiadhesion agent and evaluate its efficacy in preventing tissue adhesion in a rat
D K Kim et al.
Bioorganic & medicinal chemistry, 7(8), 1715-1725 (1999-09-11)
A series of 2-amino-9-(3-acyloxymethyl-4-alkoxycarbonyloxybut-1-yl)purin es (1-8) and 2-amino-9-(3-alkoxycarbonyl-oxymethyl-4-alkoxycarbonyloxybut -1-yl)purines (9-12) were synthesized as potential prodrugs of penciclovir. Treatment of 6-deoxypenciclovir with trimethyl orthoacetate or triethyl orthopropionate (1.2 equiv) in DMF in the presence of p-TsOH.H2O (0.1 equiv) followed by quenching
Edgar Márquez et al.
The journal of physical chemistry. A, 112(47), 12140-12142 (2008-11-01)
The gas-phase elimination kinetics of the title compounds have been examined over the temperature range of 310-369 degrees C and pressure range of 50-130 Torr. The reactions, in seasoned vessels, are homogeneous, unimolecular, and follow a first-order rate law. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service