Skip to Content
Merck
All Photos(1)

Key Documents

235857

Sigma-Aldrich

Aluminum ethoxide

97%

Synonym(s):

Aluminum triethoxide, Triethoxyaluminum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al(OC2H5)3
CAS Number:
Molecular Weight:
162.16
Beilstein:
3678970
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

97%

form

powder and chunks
solid

reaction suitability

core: aluminum

mp

154-159 °C (lit.)

SMILES string

CCO[Al](OCC)OCC

InChI

1S/3C2H5O.Al/c3*1-2-3;/h3*2H2,1H3;/q3*-1;+3

InChI key

JPUHCPXFQIXLMW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Aluminum ethoxide may be used to prepare mesoporous γ-alumina with tailormade properties. It can also be used for the growth of Al2O3 thin films by atomic layer deposition.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Skin Corr. 1B

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hailong Xiong et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 6(6), 1801543-1801543 (2019-04-03)
Mesoporous metal oxides (MMOs) have attracted comprehensive attention in many fields, including energy storage, catalysis, and separation. Current synthesis of MMOs mainly involve use of surfactants as templates to generate mesopores and organic reagents as solvents to hinder hydrolysis and
Tailored mesoporous alumina prepared from different aluminum alkoxide precursors.
May M, et al.
Journal of Porous Materials, 14(2), 159-164 (2007)
Low temperature (< 100oC) deposition of aluminum oxide thin films by ALD with O3 as oxidant.
Kim SK,et al.
Journal of the Electrochemical Society, 153(5), F69-F76 (2006)

Articles

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service