Skip to Content
Merck
All Photos(4)

Key Documents

234893

Sigma-Aldrich

Ethyl methacrylate

contains 15-20 ppm monomethyl ether hydroquinone as inhibitor, 99%

Synonym(s):

2-Methyl-2-propenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=C(CH3)COOC2H5
CAS Number:
Molecular Weight:
114.14
Beilstein:
471201
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.25

vapor density

>3.9 (vs air)

vapor pressure

15 mmHg ( 20 °C)

Assay

99%

form

liquid

autoignition temp.

771 °F

contains

15-20 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.413 (lit.)

bp

118-119 °C (lit.)

density

0.917 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOC(=O)C(C)=C

InChI

1S/C6H10O2/c1-4-8-6(7)5(2)3/h2,4H2,1,3H3

InChI key

SUPCQIBBMFXVTL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl methacrylate (EMA) is a monomer commonly used in the field of polymer industries. It is an ester of methacrylic acid and ethanol and is a colorless liquid with a characteristic odor. EMA is widely used in the production of various polymers such as poly(methyl methacrylate) (PMMA), poly(ethyl acrylate) (PEA), and ethylene-EMA (EE). These polymers find their application in various fields, including medical implant devices, packaging, paper coatings, textile coatings, emulsion polymers, and plastics.

Ethyl methacrylate is a readily polymerizable monomer used for certain types of acrylic resins. The monomethyl ether hydroxyl quinone present in it is an inhibitor that prevents polymerization.

Application

Ethyl methacrylate (EMA) can be used as a monomer in the following applications:
  • To synthesize artificial nanosized latexes of poly(styrene-co-methyl methacrylate) or poly(styrene-co-ethyl methacrylate), which are in producing drug-releasing films.
  • In the production of additive-manufactured methacrylate-based resins used in dentistry.
  • In the synthesis of a star-shaped block copolymer electrolyte for all-solid-state lithium batteries.
  • In the synthesis of a copolymer used as a matrix for semiconductor nanoparticles, which is crucial for the formation of a stable matrix for the quantum dots-copolymer composite material used in optoelectronic applications.
It can also be used to prepare copolymers of EMA on waxy maize starch and hydroxypropyl starch, the potential application of these copolymers are as excipients for compressed non-disintegrating matrix tablets. It may be used to study reactions of n-butyl acrylate and EMA with ozone in the gas phase.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

66.2 °F - closed cup

Flash Point(C)

19 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reactions of n-butyl acrylate and ethyl methacrylate with ozone in the gas phase, Computational and Theoretical Chemistry
Sun Y, et al.
Computational & Theoretical Chemistry, 1039, 33-39 (2014)
K Dharmalingam et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 69(2), 467-470 (2007-06-02)
Molecular interaction between alcohols (1-propanol, 1-butanol, s-butanol, t-butanol, 1-pentanol, 1-heptanol, 1-octanol and 1-decanol) with ethyl methacrylate has been studied in n-heptane, CCl(4) and benzene at 298K using FTIR spectroscopic and dielectric methods. The result obtained from both the methods indicates
Mengwei Pi et al.
Colloids and surfaces. B, Biointerfaces, 78(2), 193-199 (2010-03-30)
The nanoparticles composed of polystyrene core and poly[2-(diethylamino)ethyl methacrylate] (PDEA) hairy shell were used as colloidal templates for in situ silica mineralization, allowing the well-controlled synthesis of hybrid silica core-shell nanoparticles with raspberry-like morphology and hollow silica nanoparticles by subsequent
International journal of toxicology, 21 Suppl 1, 63-79 (2002-06-04)
Ethyl Methacrylate is a methacrylate ester used as a chemical additive in artificial fingernail enhancement products. These products may be applied by trained professionals or be provided directly to consumers with instructions for use. Ethyl Methacrylate readily polymerizes and rapidly
Elena Torres et al.
Polymers, 12(8) (2020-08-06)
We aimed to study the distribution of hydroxyapatite (HA) and halloysite nanotubes (HNTs) as fillers and their influence on the hydrophobic character of conventional polymers used in the biomedical field. The hydrophobic polyester poly (ε-caprolactone) (PCL) was blended with its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service