234877
Sodium iminodiacetate dibasic hydrate
98%
Synonym(s):
Iminodiacetic acid disodium salt hydrate
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Recommended Products
Assay
98%
form
solid
functional group
amine
SMILES string
O.[Na+].[Na+].[O-]C(=O)CNCC([O-])=O
InChI
1S/C4H7NO4.2Na.H2O/c6-3(7)1-5-2-4(8)9;;;/h5H,1-2H2,(H,6,7)(H,8,9);;;1H2/q;2*+1;/p-2
InChI key
YSBGCHXLMBAKPV-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Application
Sodium iminodiacetate dibasic hydrate (Iminodiacetic acid disodium salt monohydrate) has been used in the preparation of:
- different iminodiacetic acid-agarose supports, useful for selective adsorption of large proteins
- N-(o-hydroxybenzyl) iminodiacetic acid (multidentante ligand)
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
New multidentate ligands. XVII. Chelating tendencies of N-(o-hydroxybenzyl) iminodiacetic acid (H3L).
Inorganic Chemistry, 14(5), 974-978 (1975)
Journal of chromatography. A, 1055(1-2), 93-98 (2004-11-25)
A strategy to selectively adsorb large proteins on immobilized metal ion affinity chromatography supports is presented. It is based on the fact that large proteins have a large surface that permits the long distance interaction with groups placed quite far
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service