Recommended Products
Assay
99%
form
solid
refractive index
n20/D 1.486 (lit.)
bp
52-53 °C/19 mmHg (lit.)
mp
22.4 °C (lit.)
density
1.362 g/mL at 25 °C (lit.)
functional group
fluoro
SMILES string
Oc1ccc(F)cc1F
InChI
1S/C6H4F2O/c7-4-1-2-6(9)5(8)3-4/h1-3,9H
InChI key
NVWVWEWVLBKPSM-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
2,4-Difluorophenol has been used:
- as starting reagent during the synthesis of 4,6-difluoro-5,7-dihydroxytryptamine
- in the synthesis of 3-carboxy-6,8-difluoro-7-hydroxycoumarin (Pacific Blue), fluorinated fluorescent dye
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
134.6 °F - closed cup
Flash Point(C)
57 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Genetically determined differences in noradrenergic input to the brain cortex: a histochemical and biochemical study in two inbred strains of mice.
Neuroscience, 4(7), 877-888 (1979-01-01)
Bioorganic & medicinal chemistry letters, 8(22), 3107-3110 (1999-01-05)
Two new fluorinated fluorescent dyes, 6,8-difluoro-7-hydroxy-4-methylcoumarin (Marina Blue) and 3-carboxy-6,8-difluoro-7-hydroxycoumarin (Pacific Blue), exhibit excellent photophysical properties among a series of novel fluorinated 7-hydroxycoumarins. Most of these fluorinated coumarins have quantum yields (0.63 to 0.89) equal to or higher than that
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service