Skip to Content
Merck
All Photos(1)

Key Documents

232270

Sigma-Aldrich

3-Amino-5-methylisoxazole

≥97%

Synonym(s):

5-Methyl-3-isoxazolamine, NSC 159134

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N2O
CAS Number:
Molecular Weight:
98.10
Beilstein:
108110
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥97%

form

solid

mp

59-61 °C (lit.)

SMILES string

Cc1cc(N)no1

InChI

1S/C4H6N2O/c1-3-2-4(5)6-7-3/h2H,1H3,(H2,5,6)

InChI key

FKPXGNGUVSHWQQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Amino-5-methylisoxazole is the major intermediate formed during sulfamethoxazole biodegradation by Pseudomonas psychrophila strain HA-4[1]. It is the intermediate formed during the photocatalytic degradation of sulfamethoxazole (SMX)[2].

Application

3-Amino-5-methylisoxazole was used in synthesis of:
  • naphtho[1,2-e][1,3]oxazines[3]
  • series of 1-aryl-4-methyl-3,6-bis-(5-methylisoxazol-3-yl)-2-thioxo-2,3,6,10b-tetrahydro-1H-pyrimido[5,4-c]quinolin-5-ones, having potential mosquito larvicidal activity[4]
  • hydroxylamines of sulfadiazine and sulfamethoxazole[5]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yan Li et al.
The Science of the total environment, 633, 1217-1226 (2018-05-16)
Sulfamethoxazole (SMX) is a sulfonamide antibiotic, widely used as curative and preventive drug for human, animal, and aquaculture bacterial infections. Its residues have been ubiquitously detected in the surface waters and sediments. In the present study, SMX dissipation and kinetics
Shiyuan Ding et al.
Journal of hazardous materials, 262, 812-818 (2013-10-22)
Photocatalytic degradation of sulfamethoxazole (SMX) was investigated using Bi2O3/Bi2O2CO3/Sr6Bi2O9 (BSO) photocatalyst under visible light (>420 nm) irradiation. The photochemical degradation of SMX followed pseudo-first-order kinetics. The reaction kinetics was determined as a function of initial SMX concentrations (5-20 mg L(-1))
Lu Wang et al.
Water research, 88, 322-328 (2015-10-30)
Sulphamethoxazole (SMX) is extensively used in humans and livestock, but its appearance in natural water raises environmental concerns. This study demonstrated that SMX and its degradation product, 3-amino-5-methylisoxazole (3A5MI), could be effectively degraded in microbial fuel cell (MFC) reactors. Approximately
Bianca M Souza et al.
Environmental science and pollution research international, 24(7), 6195-6204 (2015-11-12)
The present study aims to assess the removal of 3-amino-5-methylisoxazole (AMI), a recalcitrant by-product resulting from the biological breakdown of some pharmaceuticals, applying a solar photo-Fenton process assisted by ferrioxalate complexes (SPFF) (Fe
Mehdi Shafiee et al.
Molecular diversity, 16(4), 727-735 (2012-10-24)
An expeditious, straightforward and efficient synthesis of diversely naphtho[1,2-e][1,3]oxazines via one-pot condensation reaction of β- naphthol, 3-amino-5-methylisoxazole and arylaldehydes catalyzed by bismuth(III) trifluoromethanesulfonate is described. The reaction preferentially afforded 1,3-trans oxazines.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service