Skip to Content
Merck
All Photos(2)

Key Documents

230715

Sigma-Aldrich

n-Butyllithium solution

11.0 M in hexanes

Synonym(s):

n-BuLi, Butyl lithium, Butyllithium solution, Lithium-1-butanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3Li
CAS Number:
Molecular Weight:
64.06
Beilstein:
1209227
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

concentration

11.0 M in hexanes

density

0.761 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

[Li]CCCC

InChI

1S/C4H9.Li/c1-3-4-2;/h1,3-4H2,2H3;

InChI key

MZRVEZGGRBJDDB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

n-Butyllithium (n-BuLi) is an organolithium reagent commonly used as a strong base to form corresponding lithium salts by the deprotonation of nitrogen, oxygen, phosphorus, and carbon acids. Heterocyclic compounds, such as furans, thiophenes, oxazoles, pyrroles, etc, can be lithiated α to the ring heteroatom using n?BuLi. These lithiated salts reacting in situ with alkyl halides to obtain useful organic compounds by the formation of the C-C bond.

Application

n-BuLi can be used as a reagent for lithium-halogen exchange reactions. Synthesis of various other useful reagents such as lithium diisopropylamide (LDA) and diphenylphosphine is prepared by in situ reaction with n-BuLi . In the polymerization of dienes, it can be employed as an initiator.

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Repr. 2 - Skin Corr. 1B - STOT RE 1 Inhalation - STOT SE 3 - Water-react 1

Target Organs

Central nervous system, Nervous system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

-7.6 °F - closed cup

Flash Point(C)

-22 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

n-Butyllithium
Ovaska TV, et al.
Encyclopedia of Reagents for Organic Synthesis, Second Edition (2001)
Preparation, Properties, and Safe Handling of Commercial Organolithiums: Alkyllithiums, Lithium sec-Organoamides, and Lithium Alkoxides.
Rathman, Terry `Li? and Schwindeman, James A
Organic Process Research & Development, 18(10), 1192-1210 (2014)
n-Butyllithium.
eEROS (Encyclopedia of Reagents for Organic Synthesis) (2001)
Microstructure-thermal property relationship of high trans-1, 4-poly (butadiene) produced by anionic polymerization of 1, 3-butadiene using an initiator composed of alkyl aluminum, n-butyl lithium, and barium alkoxide
Benvenuta-Tapia JJ, et al.
Polym. Eng. Sci., 49(1), 1-10 (2009)
Diphenylphosphine.
Bianco, VD et al.
Inorganic Syntheses, 16(10), 161-163 (1976)

Articles

Transformative reagents enable selective conversions within molecules containing sensitive functionalities under mild reactions.

Transformative reagents enable selective conversions within molecules containing sensitive functionalities under mild reactions.

Transformative reagents enable selective conversions within molecules containing sensitive functionalities under mild reactions.

Transformative reagents enable selective conversions within molecules containing sensitive functionalities under mild reactions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service