Skip to Content
Merck
All Photos(1)

Key Documents

224766

Sigma-Aldrich

N-Methylcaprolactam

99%

Synonym(s):

Hexahydro-1-methyl-2H-azepin-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H13NO
CAS Number:
Molecular Weight:
127.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.484 (lit.)

bp

106-108 °C/6 mmHg (lit.)

density

0.991 g/mL at 25 °C (lit.)

SMILES string

CN1CCCCCC1=O

InChI

1S/C7H13NO/c1-8-6-4-2-3-5-7(8)9/h2-6H2,1H3

InChI key

ZWXPDGCFMMFNRW-UHFFFAOYSA-N

Application

N-Methylcaprolactam was used to study the inhibition mechanism of poly-[N-vinylcaprolactam] (PVCAP) and its related compounds against hydrate formation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

217.4 °F - closed cup

Flash Point(C)

103 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michika Ohtake et al.
The journal of physical chemistry. B, 109(35), 16879-16885 (2006-07-21)
Poly-[N-vinylcaprolactam] (PVCAP) and its related compounds are specific polymeric compounds for inhibiting hydrate formation. To clarify the inhibition mechanism of these compounds on hydrate nucleation at the molecular level, we measured the mass spectra of clusters generated from the fragmentation
Ilnaz T Rakipov et al.
Molecules (Basel, Switzerland), 26(5) (2021-04-04)
In the present work, the thermochemistry of solution, solvation, and hydrogen bonding of cyclic amides in proton acceptor (B) and proton donor (RXH) solvents were studied. The infinite dilution solution enthalpies of δ-valerolactam, N-methylvalerolactam, ε-caprolactam, and N-methylcaprolactam were measured at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service